Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XID | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336437.1 | NP_001324213.1 | 15 - 16 | 2232 | 76 | |
NM_001336439.1 | NP_001324209.1 | 30 - 31 | 3697 | 76 | ||
NM_001336440.1 | NP_001324214.1 | 30 - 31 | 3697 | 76 | ||
NM_128883.3 | NP_180882.2 | 29 - 30 | 3760 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |