Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NGA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116062.3 | NP_191756.1 | 1 | 1270 | 66 | |
GenBank mRNA | BT025866.1 | - | 1 | 764 | 66 | |
FJ873732.1 | - | 1 | 1130 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |