Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANTR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116261.4 | NP_567175.2 | 2 - 3 | 366 | 69 | |
GenBank mRNA | AK118390.1 | - | 2 - 3 | 241 | 69 | |
BT009663.1 | - | 2 - 3 | 120 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |