Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340369.1 | NP_001329795.1 | 23 - 24 | 2813 | 106 | |
NM_116459.4 | NP_567226.1 | 23 - 24 | 2875 | 106 | ||
NM_178956.4 | NP_849287.2 | 23 - 24 | 2869 | 106 | ||
NM_202772.2 | NP_974501.1 | 22 - 23 | 2952 | 106 | ||
GenBank mRNA | AF225702.1 | - | 23 - 24 | 2719 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |