Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116461.4 | NP_192137.1 | 9 - 10 | 2460 | 68 | |
GenBank mRNA | AY051001.1 | - | 9 - 10 | 2234 | 68 | |
AY056784.1 | - | 9 - 10 | 2259 | 68 | ||
AY142511.1 | - | 9 - 10 | 2020 | 68 | ||
KJ138769.1 | - | 9 - 10 | 2020 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |