Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IDD12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340394.1 | NP_001328388.1 | 1 - 2 | 318 | 111 | |
NM_116501.2 | NP_192176.1 | 1 - 2 | 183 | 111 | ||
GenBank mRNA | AB493672.1 | - | 1 - 2 | 183 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |