Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116594.5 | NP_567258.1 | 17 - 18 | 2077 | 95 | |
GenBank mRNA | AF360372.1 | - | 17 - 18 | 1753 | 95 | |
AK316725.1 | - | 17 - 18 | 1794 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |