Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SULTR1;1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116931.3 | NP_192602.1 | 8 - 9 | 1241 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |