Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084247.2 | NP_001077716.1 | 4 - 5 | 732 | 122 | |
NM_104251.4 | NP_175777.1 | 4 - 5 | 655 | 122 | ||
GenBank mRNA | AK227057.1 | - | 4 - 5 | 408 | 122 | |
AY518291.1 | - | 4 - 5 | 436 | 122 | ||
BT010600.1 | - | 4 - 5 | 366 | 122 | ||
FJ708659.1 | - | 4 - 5 | 366 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |