Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XIE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333655.1 | NP_001320715.1 | 31 - 32 | 4296 | 67 | |
NM_104334.2 | NP_175858.1 | 30 - 31 | 4020 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |