Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSAG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104443.2 | NP_175963.1 | 1 | 632 | 145 | |
GenBank mRNA | AF324710.2 | - | 1 | 632 | 145 | |
AF326870.1 | - | 1 | 632 | 145 | ||
AF378859.1 | - | 1 | 634 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |