Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333724.1 | NP_001320656.1 | 5 - 6 | 623 | 79 | |
NM_104448.4 | NP_175968.4 | 5 - 6 | 668 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |