Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BGLU45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198358.1 | NP_001185287.1 | 7 - 8 | 794 | 70 | |
NM_001334002.1 | NP_001319288.1 | 7 - 8 | 888 | 70 | ||
NM_001334003.1 | NP_001322277.1 | 7 - 8 | 792 | 70 | ||
NM_001334004.1 | NP_001322278.1 | 4 - 5 | 585 | 70 | ||
NM_104863.3 | NP_176374.1 | 7 - 8 | 888 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |