Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SKU5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203775.2 | NP_001190704.1 | 8 - 9 | 1935 | 71 | |
NM_117312.4 | NP_192979.1 | 8 - 9 | 1917 | 71 | ||
GenBank mRNA | AF439406.1 | - | 8 - 9 | 1668 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |