Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340905.1 | NP_001319933.1 | 3 - 4 | 753 | 65 | |
NM_001340906.1 | NP_001329589.1 | 3 - 4 | 753 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |