Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340905.1 | NP_001319933.1 | 7 - 8 | 3037 | 79 | |
NM_001340906.1 | NP_001329589.1 | 7 - 8 | 2980 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |