Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRPE7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340952.1 | NP_001329853.1 | 2 | 998 | 105 | |
NM_001340953.1 | NP_001329854.1 | 2 | 675 | 105 | ||
NM_117548.4 | NP_193202.2 | 2 | 834 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |