Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117706.3 | NP_193348.1 | 28 - 29 | 3098 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |