Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGLYAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117809.6 | NP_193438.2 | 9 - 10 | 816 | 118 | |
GenBank mRNA | AK118019.1 | - | 9 - 10 | 701 | 118 | |
GQ303359.1 | - | 9 - 10 | 655 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |