Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IP5PII | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341251.1 | NP_001319979.1 | 7 - 8 | 1816 | 99 | |
NM_117911.4 | NP_567547.1 | 7 - 8 | 1907 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |