Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117980.5 | NP_193599.3 | 9 - 10 | 2009 | 126 | |
GenBank mRNA | FJ708745.1 | - | 8 - 9 | 1294 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |