Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343719.1 | NP_001332528.1 | 3 - 4 | 590 | 106 | |
NM_147892.4 | NP_680197.2 | 3 - 4 | 590 | 106 | ||
GenBank mRNA | BT022108.1 | - | 3 - 4 | 511 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |