Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KUP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105698.5 | NP_177187.2 | 2 - 3 | 927 | 108 | |
GenBank mRNA | AY062542.1 | - | 2 - 3 | 622 | 108 | |
AY093328.1 | - | 2 - 3 | 530 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |