Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCB26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334463.1 | NP_001321943.1 | 17 - 18 | 2159 | 127 | |
NM_105729.6 | NP_177218.3 | 17 - 18 | 2208 | 127 | ||
GenBank mRNA | AK228750.1 | - | 17 - 18 | 2146 | 127 | |
AY078409.1 | - | 17 - 18 | 2021 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |