Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALAAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084342.1 | NP_001077811.1 | 10 - 11 | 1222 | 108 | |
NM_001198451.1 | NP_001185380.1 | 10 - 11 | 1271 | 108 | ||
NM_105892.5 | NP_565040.2 | 10 - 11 | 1289 | 108 | ||
GenBank mRNA | AF275371.1 | - | 10 - 11 | 1203 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |