Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NF-YC12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123174.5 | NP_198630.2 | 3 - 4 | 577 | 59 | |
GenBank mRNA | BT021919.1 | - | 3 - 4 | 325 | 59 | |
BT025775.1 | - | 2 - 3 | 192 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |