Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZIFL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339105.1 | NP_001326438.1 | 5 - 6 | 570 | 106 | |
NM_001339106.1 | NP_001326437.1 | 5 - 6 | 566 | 106 | ||
NM_114247.4 | NP_189965.2 | 5 - 6 | 570 | 106 | ||
NM_202653.1 | NP_974382.1 | 5 - 6 | 505 | 106 | ||
NM_202654.1 | NP_974383.1 | 5 - 6 | 505 | 106 | ||
GenBank mRNA | BT030378.1 | - | 5 - 6 | 446 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |