Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339141.1 | NP_001326422.1 | 3 - 4 | 2195 | 105 | |
NM_001339142.1 | NP_001326421.1 | 3 - 4 | 2258 | 105 | ||
NM_001339143.1 | NP_001326419.1 | 3 - 4 | 2258 | 105 | ||
NM_001339144.1 | NP_001326420.1 | 4 - 5 | 2016 | 105 | ||
NM_114316.3 | NP_190034.2 | 4 - 5 | 2079 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |