Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100089.3 | NP_563645.1 | 4 - 5 | 654 | 97 | |
NM_179245.3 | NP_849576.1 | 4 - 5 | 654 | 97 | ||
NM_202010.3 | NP_973739.1 | 4 - 5 | 654 | 97 | ||
GenBank mRNA | AF063852.2 | - | 4 - 5 | 458 | 97 | |
AF372934.1 | - | 4 - 5 | 489 | 97 | ||
AF395065.1 | - | 4 - 5 | 417 | 97 | ||
AF395066.1 | - | 4 - 5 | 417 | 97 | ||
AY087971.1 | - | 4 - 5 | 492 | 97 | ||
AY133518.1 | - | 4 - 5 | 414 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |