Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLDP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111436.6 | NP_187214.2 | 14 - 15 | 2912 | 90 | |
GenBank mRNA | AM182458.1 | - | 14 - 15 | 2838 | 90 | |
AY305003.1 | - | 14 - 15 | 2785 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |