Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111616.3 | NP_566305.1 | 5 - 6 | 692 | 97 | |
GenBank mRNA | AY042807.1 | - | 5 - 6 | 692 | 97 | |
AY064647.1 | - | 5 - 6 | 444 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |