Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341861.1 | NP_001327998.1 | 7 - 8 | 1097 | 82 | |
NM_118890.2 | NP_194485.1 | 7 - 8 | 1097 | 82 | ||
GenBank mRNA | KJ676985.1 | - | 7 - 8 | 1057 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |