Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036690.1 | NP_001031767.1 | 8 - 9 | 863 | 110 | |
NM_001203956.1 | NP_001190885.1 | 8 - 9 | 820 | 110 | ||
NM_119351.3 | NP_194927.1 | 7 - 8 | 882 | 110 | ||
NM_179152.3 | NP_849483.1 | 8 - 9 | 895 | 110 | ||
GenBank mRNA | AK316993.1 | - | 8 - 9 | 748 | 110 | |
AK317084.1 | - | 7 - 8 | 883 | 110 | ||
AY054660.1 | - | 8 - 9 | 865 | 110 | ||
AY081506.1 | - | 7 - 8 | 655 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |