Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085022.1 | NP_001078491.1 | 5 - 6 | 1325 | 101 | |
NM_119611.5 | NP_195172.1 | 5 - 6 | 1454 | 101 | ||
NM_179163.3 | NP_849494.2 | 4 - 5 | 1577 | 101 | ||
NM_202954.3 | NP_974683.1 | 5 - 6 | 1351 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |