Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BGAL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119799.4 | NP_568001.1 | 11 - 12 | 1583 | 144 | |
NM_179175.2 | NP_849506.1 | 11 - 12 | 1378 | 144 | ||
GenBank mRNA | AJ270299.1 | - | 11 - 12 | 1300 | 144 | |
AY056285.1 | - | 11 - 12 | 1359 | 144 | ||
AY091432.1 | - | 11 - 12 | 1260 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |