Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160815.2 | NP_001154287.1 | 1 - 2 | 645 | 118 | |
NM_001203993.1 | NP_001190922.1 | 2 - 3 | 275 | 118 | ||
NM_001203994.1 | NP_001190923.1 | 2 - 3 | 275 | 118 | ||
NM_001342339.1 | NP_001328125.1 | 2 - 3 | 387 | 118 | ||
NM_001342340.1 | NP_001328126.1 | 2 - 3 | 323 | 118 | ||
NM_001342341.1 | NP_001328129.1 | 2 - 3 | 530 | 118 | ||
NM_001342342.1 | NP_001328130.1 | 1 - 2 | 821 | 118 | ||
NM_001342343.1 | NP_001328127.1 | 2 - 3 | 555 | 118 | ||
NM_001342344.1 | NP_001328128.1 | 2 - 3 | 523 | 118 | ||
NM_119696.5 | NP_195256.3 | 2 - 3 | 388 | 118 | ||
NM_179234.2 | NP_849565.1 | 2 - 3 | 357 | 118 | ||
GenBank mRNA | AJ532570.1 | - | 1 - 2 | 81 | 118 | |
AY094465.1 | - | 2 - 3 | 289 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |