Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331755.1 | NP_001322007.1 | 3 - 4 | 1635 | 89 | |
NM_100680.4 | NP_563806.1 | 3 - 4 | 1635 | 89 | ||
NM_179277.1 | NP_849608.1 | 2 - 3 | 1367 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |