Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALIS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112110.4 | NP_566435.1 | 7 - 8 | 1432 | 71 | |
GenBank mRNA | AF410309.1 | - | 7 - 8 | 1061 | 71 | |
AY045904.1 | - | 7 - 8 | 1137 | 71 | ||
AY087607.1 | - | 7 - 8 | 1226 | 71 | ||
AY091210.1 | - | 7 - 8 | 1002 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |