Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OVA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112195.3 | NP_187958.1 | 13 - 14 | 1769 | 66 | |
GenBank mRNA | BT002317.1 | - | 13 - 14 | 1721 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |