Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSP60-3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112240.4 | NP_566466.1 | 15 - 16 | 1745 | 77 | |
GenBank mRNA | AY094453.1 | - | 15 - 16 | 1659 | 77 | |
BT015923.1 | - | 15 - 16 | 1543 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |