Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342099.1 | NP_001328023.1 | 1 - 2 | 272 | 126 | |
NM_119310.4 | NP_567880.1 | 1 - 2 | 272 | 126 | ||
GenBank mRNA | BT003891.1 | - | 1 - 2 | 272 | 126 | |
BT005002.1 | - | 1 - 2 | 71 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |