Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LUG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203962.1 | NP_001190891.1 | 11 - 12 | 2403 | 78 | |
NM_119407.3 | NP_567896.1 | 12 - 13 | 2345 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |