Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KUP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342219.1 | NP_001329920.1 | 2 - 3 | 1081 | 65 | |
NM_119508.4 | NP_195079.2 | 3 - 4 | 1025 | 65 | ||
GenBank mRNA | AY099556.1 | - | 3 - 4 | 855 | 65 | |
BT010357.1 | - | 3 - 4 | 770 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |