Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DML3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125640.1 | NP_001119112.1 | 5 - 6 | 1216 | 121 | |
NM_001342250.1 | NP_001329149.1 | 6 - 7 | 1443 | 121 | ||
NM_001342251.1 | NP_001329150.1 | 3 - 4 | 1468 | 121 | ||
NM_001342252.1 | NP_001329148.1 | 4 - 5 | 1316 | 121 | ||
NM_119567.4 | NP_195132.3 | 5 - 6 | 1370 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |