Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCS16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119589.6 | NP_195151.1 | 1 | 802 | 103 | |
GenBank mRNA | AK229262.1 | - | 1 | 682 | 103 | |
BT030458.1 | - | 1 | 653 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |