Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342287.1 | NP_001329504.1 | 5 - 6 | 1200 | 69 | |
NM_119614.6 | NP_195175.1 | 5 - 6 | 1159 | 69 | ||
GenBank mRNA | AB014759.1 | - | 5 - 6 | 1036 | 69 | |
BT029161.1 | - | 5 - 6 | 981 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |