Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106591.2 | NP_565209.1 | 3 - 4 | 547 | 79 | |
NM_179574.3 | NP_849905.1 | 4 - 5 | 532 | 79 | ||
GenBank mRNA | AK316719.1 | - | 4 - 5 | 458 | 79 | |
AY045662.1 | - | 3 - 4 | 545 | 79 | ||
AY143841.1 | - | 3 - 4 | 193 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |