Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLASP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179666.5 | NP_849997.2 | 12 - 13 | 1793 | 76 | |
GenBank mRNA | AY091025.1 | - | 12 - 13 | 1686 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |