Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLASP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179666.5 | NP_849997.2 | 9 - 10 | 1493 | 84 | |
GenBank mRNA | AY091025.1 | - | 9 - 10 | 1386 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |