Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179672.4 | NP_850003.1 | 2 - 3 | 468 | 72 | |
GenBank mRNA | AY063934.1 | - | 2 - 3 | 393 | 72 | |
AY096480.1 | - | 2 - 3 | 331 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |